N-Boc-2-methyl-D-serine methyl ester 97%
Lieferant: Thermo Scientific
H52421.03
H52421.03EA
455
EUR
H52421.03
N-Boc-2-methyl-D-serine methyl ester 97%
N-Boc-2-methyl-D-serine methyl ester
Purity: 97%
CAS No.: 188476-33-7
Molecular Formula: C10H19NO5
Formula Weight: 233.26
MDL No.: MFCD06797553
Formel:
C₁₀H₁₉NO₅ Molecular Weight: 233,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797553 CAS-Nummer: 188476-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...