N-Boc-3-dimethylamino-D-alanine 97%
Lieferant: Thermo Fisher Scientific
H52433.03
H52433.03EA
239
EUR
H52433.03
N-Boc-3-dimethylamino-D-alanine 97%
N-Boc-3-dimethylamino-D-alanine
Purity: 97%
CAS No.: 110755-32-3
Molecular Formula: C10H20N2O4
Formula Weight: 232.28
Formel:
C₁₀H₂₀N₂O₄ Molecular Weight: 232.28 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
110755-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...