2-(Brommethyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H52467.03 H52467.06
H52467.03EA 96.6 EUR
H52467.03 H52467.06
2-(Brommethyl)phenylboronsäure ≥97%
2-(Brommethyl)phenylboronsäure

Purity: 97% CAS No.: 91983-14-1 Molecular Formula: C7H8BBrO2 Formula Weight: 214.85 Melting Point: 145-148° MDL No.: MFCD01318941

Formel: C₇H₈BBrO₂
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318941
CAS-Nummer: 91983-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR