2-(2,2,2-Trifluorethoxy)phenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H52471.MD
H52471.MDEA
148
EUR
H52471.MD
2-(2,2,2-Trifluorethoxy)phenylboronsäure ≥98%
2-(2,2,2-Trifluorethoxy)phenylboronsäure
Purity: 98%
CAS No.: 957060-90-1
Molecular Formula: C8H8BF3O3
Formula Weight: 219.95
Storage Temperature: 2°C to 8°C
Melting Point: 125-128°
MDL No.: MFCD09027258
Formel:
C₈H₈BF₃O₃ Schmelzpunkt: 125…128 °C |
MDL:
MFCD09027258 CAS-Nummer: 957060-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...