3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(3-((2-(Dimethylamino)ethyl)carbamoyl)phenyl)boronic acid
H52498.03
H52498.03EA
302
EUR
H52498.03
3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid 96%
3-[2-(Dimethylamino)ethylcarbamoyl]benzeneboronic acid
Purity: 96%
CAS No.: 850567-31-6
Molecular Formula: C11H17BN2O3
Formula Weight: 236.08
Melting Point: 42-45°
MDL No.: MFCD04115706
Formel:
C₁₁H₁₇BN₂O₃ Molecular Weight: 236,08 g/mol Schmelzpunkt: 42…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115706 CAS-Nummer: 850567-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...