3-Fluor-(4-methoxycarbonyl)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H52510.03
H52510.06
H52510.03EA
127
EUR
H52510.03
H52510.06
3-Fluor-(4-methoxycarbonyl)phenylboronsäure ≥97%
3-Fluor-(4-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BFO₄ Schmelzpunkt: 180…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115665 CAS-Nummer: 505083-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...