5-Methyl-2-pyridinboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (5-Methylpyridin-2-yl)boronic acid

H52517.MD
H52517.MDEA 172 EUR
H52517.MD
5-Methyl-2-pyridinboronsäure 95%
5-Methyl-2-pyridineboronic acid

Purity: 95% CAS No.: 372963-49-0 Molecular Formula: C6H8BNO2 Formula Weight: 136.95 Storage Temperature: -30°C to -10°C Melting Point: 86-90° MDL No.: MFCD00674179

Formel: C₆H₈BNO₂
Molecular Weight: 136,95 g/mol
Schmelzpunkt: 86…90 °C
MDL: MFCD00674179
CAS-Nummer: 372963-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR