1-Cyclopentenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Cyclopent-1-en-1-ylboronic acid

H52531.MD
H52531.MDEA 261 EUR
H52531.MD
1-Cyclopentenylboronsäure ≥97%
1-Cyclopentenylboronsäure

Purity: 97% CAS No.: 850036-28-1 Molecular Formula: C5H9BO2 Formula Weight: 111.94 Storage Temperature: -30°C to -10°C Melting Point: 134-140° MDL No.: MFCD02179496

Formel: C₅H₉BO₂
Molecular Weight: 111,94 g/mol
Schmelzpunkt: 134…140 °C
MDL: MFCD02179496
CAS-Nummer: 850036-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR