(R)-1-Boc-3-hydroxypiperidin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H52556.03
H52556.06
H52556.03EA
69.2
EUR
H52556.03
H52556.06
(R)-1-Boc-3-hydroxypiperidin ≥97%
(R)-1-Boc-3-hydroxypiperidin
Purity: 97%
CAS No.: 143900-43-0
Molecular Formula: C10H19NO3
Formula Weight: 201.27
Flash Point: 104°(219°F)
Melting Point: 46-50°
Optical Rotation: -23° (c=5 in methanol)
MDL No.: MFCD04115306
Formel:
C₁₀H₁₉NO₃ Schmelzpunkt: 46…50 °C Flammpunkt: 104 °C (219 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115306 CAS-Nummer: 143900-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...