1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Hydroxy-1,2,3,6-tetrahydropyridine, N-BOC protected, tert-Butyl 5-hydroxy-5,6-dihydropyridine-1(2H)-carboxylate

H52577.03
H52577.03EA 181 EUR
H52577.03
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine 97%
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine

Purity: 97% CAS No.: 224779-27-5 Molecular Formula: C10H17NO3 Formula Weight: 199.25 MDL No.: MFCD04972246

Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972246
CAS-Nummer: 224779-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR