trans-1-Hexen-1-ylboronic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(E)-Hex-1-en-1-ylboronic acid
H52613.03
H52613.03EA
146
EUR
H52613.03
trans-1-Hexen-1-ylboronic acid 98%
trans-1-Hexen-1-ylboronic acid
Purity: 98%
CAS No.: 42599-18-8
Molecular Formula: C6H13BO2
Formula Weight: 127.98
Storage Temperature: -30°C to -10°C
Melting Point: 62-63°
MDL No.: MFCD01074553
Formel:
C₆H₁₃BO₂ Molecular Weight: 127,98 g/mol Schmelzpunkt: 62…63 °C |
MDL:
MFCD01074553 CAS-Nummer: 42599-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...