trans-3,3-Dimethyl-1-butenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-tert-Butyl-trans-vinylboronic acid, (E)-(3,3-Dimethylbut-1-en-1-yl)boronic acid

H52627.MD
H52627.MDEA 89.6 EUR
H52627.MD
trans-3,3-Dimethyl-1-butenylboronsäure ≥97%
trans-3,3-Dimethyl-1-butenylboronic acid

Purity: 97% CAS No.: 86595-37-1 Molecular Formula: C6H13BO2 Formula Weight: 127.98 Melting Point: 78-80° MDL No.: MFCD01074624

Formel: C₆H₁₃BO₂
Molecular Weight: 127,98 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01074624
CAS-Nummer: 86595-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR