3-Boronobenzolsulfonamid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Aminosulphonyl)benzeneboronic acid, 3-Sulphamoylbenzeneboronic acid
H52628.MD
H52628.MDEA
412
EUR
H52628.MD
3-Boronobenzolsulfonamid ≥97%
3-Boronobenzolsulfonamid
Purity: 97%
CAS No.: 850568-74-0
Molecular Formula: C6H8BNO4S
Formula Weight: 201.00
Storage Temperature: 2°C to 8°C
Melting Point: 260-266°
MDL No.: MFCD06659840
Formel:
C₆H₈BNO₄S Molecular Weight: 201,00 g/mol Schmelzpunkt: 260…266 °C |
MDL:
MFCD06659840 CAS-Nummer: 850568-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...