4-Methoxy-3-(trifluormethyl)phenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Methoxy-3-(trifluoromethyl)phenyl)boronic acid, 4-Methoxy-3-trifluoromethylphenylboronic acid, 4-Methoxy-3-(trifluoromethyl)benzeneboronic acid

H52660.03
H52660.03EA 216 EUR
H52660.03
4-Methoxy-3-(trifluormethyl)phenylboronsäure ≥98%
4-Methoxy-3-(trifluormethyl)phenylboronsäure

Purity: 98% CAS No.: 149507-36-8 Molecular Formula: C8H8BF3O3 Formula Weight: 219.95 Melting Point: 198-202° MDL No.: MFCD07363790

Formel: C₈H₈BF₃O₃
Molecular Weight: 219,95 g/mol
Schmelzpunkt: 198…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363790
CAS-Nummer: 149507-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR