(3-Methylsulphonylaminomethyl)benzeneboronic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(3-(Methylsulphonamidomethyl)phenyl)boronic acid
H52705.03
H52705.03EA
196
EUR
H52705.03
(3-Methylsulphonylaminomethyl)benzeneboronic acid 98%
(3-Methylsulphonylaminomethyl)benzeneboronic acid
Purity: 98%
CAS No.: 850568-39-7
Molecular Formula: C8H12BNO4S
Formula Weight: 229.06
Melting Point: 246-248°
MDL No.: MFCD06659817
Formel:
C₈H₁₂BNO₄S Molecular Weight: 229,06 g/mol Schmelzpunkt: 246…248 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659817 CAS-Nummer: 850568-39-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...