3-Chlor-4-(methylcarbamoyl)phenylboronsäure 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Chloro-4-(N-methylcarbamoyl)phenylboronic acid, (3-Chloro-4-(methylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(methylcarbamoyl)benzeneboronic acid

H52721.MD
H52721.MDEA 119 EUR
H52721.MD
3-Chlor-4-(methylcarbamoyl)phenylboronsäure 96%
3-Chlor-4-(methylcarbamoyl)phenylboronsäure

Purity: 96% CAS No.: 850589-39-8 Molecular Formula: C8H9BClNO3 Formula Weight: 213.43 Melting Point: 164-170° MDL No.: MFCD07363766

Formel: C₈H₉BClNO₃
Molecular Weight: 213,43 g/mol
Schmelzpunkt: 164…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363766
CAS-Nummer: 850589-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR