Boc-cis-4-amino-L-proline methyl ester hydrochloride salt 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Methyl (2S,4S)-1-Boc-4-aminopyrrolidine-2-carboxylate hydrochloride
H52731.03
H52731.06
H52731.03EA
190
EUR
H52731.03
H52731.06
Boc-cis-4-amino-L-proline methyl ester hydrochloride salt 97%
Boc-cis-4-amino-L-proline methyl ester hydrochloride salt
Purity: 97%
CAS No.: 171110-72-8
Molecular Formula: C11H20N2O4.HCl
Formula Weight: 280.75
Melting Point: ca 185° dec.
Optical Rotation: -40° (c=2 in methanol)
MDL No.: MFCD03787927
Formel:
C₁₁H₂₁ClN₂O₄ Schmelzpunkt: ∼ 185 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787927 CAS-Nummer: 171110-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...