trans-N-Boc-4-cyano-L-proline methyl ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-tert-Butyl 2-methyl (2S,4R)-4-cyanopyrrolidine-1,2-dicarboxylate

H52733.03
H52733.03EA 642 EUR
H52733.03
trans-N-Boc-4-cyano-L-proline methyl ester 97%
trans-N-Boc-4-cyano-L-proline methyl ester

Purity: 97% CAS No.: 194163-91-2 Molecular Formula: C12H18N2O4 Formula Weight: 254.29 MDL No.: MFCD03094915

Formel: C₁₂H₁₈N₂O₄
Molecular Weight: 254.286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094915
CAS-Nummer: 194163-91-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR