(S)-1-Boc-2-ethylpiperazin ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2S)-2-Ethylpiperazine, N1-BOC protected, (S)-tert-Butyl 2-ethylpiperazine-1-carboxylate

H52738.MD
H52738.MDEA 167 EUR
H52738.MD
(S)-1-Boc-2-ethylpiperazin ≥97%
(S)-1-Boc-2-ethylpiperazin

Purity: 97% CAS No.: 325145-35-5 Molecular Formula: C11H22N2O2 Formula Weight: 214.30 MDL No.: MFCD07772100

Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772100
CAS-Nummer: 325145-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR