Methyl-2,2-difluor-2-(fluorsulfonyl)acetat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H52745.06
H52745.14
H52745.06EA
59.5
EUR
H52745.06
H52745.14
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat ≥97%
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Purity: 97%
CAS No.: 680-15-9
Molecular Formula: C3H3F3O4S
Formula Weight: 192.11
Flash Point: 47°(117°F)
Refractive Index: 1.351
Boiling Point: 117-118°
Beilstein Reference: 1812896
UN No.: 2920
MDL No.: MFCD00144316
Formel:
C₃H₃F₃O₄S Siedepunkt: 117…118 °C Dichte: 1,509 Flammpunkt: 47 °C (117 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144316 CAS-Nummer: 680-15-9 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...