3-Fluor-2-methylphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3-Fluoro-2-methylphenyl)boronic acid, 3-Fluoro-2-methylbenzeneboronic acid

H52752.03
H52752.03EA 236 EUR
H52752.03
3-Fluor-2-methylphenylboronsäure ≥98%
3-Fluor-2-methylphenylboronsäure

Purity: 98% CAS No.: 163517-61-1 Molecular Formula: C7H8BFO2 Formula Weight: 153.95 Melting Point: 142-144° MDL No.: MFCD07368679

Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368679
CAS-Nummer: 163517-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR