2-(Dimethylcarbamoyl)phenylboronsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Dimethylcarbamoyl)benzeneboronic acid

H52765.MD H52765.03
H52765.MDEA 75 EUR
H52765.MD H52765.03
2-(Dimethylcarbamoyl)phenylboronsäure 95%
2-(Dimethylcarbamoyl)phenylboronsäure

Purity: 95% CAS No.: 874219-16-6 Molecular Formula: C9H12BNO3 Formula Weight: 193.01 Melting Point: 108-110° MDL No.: MFCD03425959

Formel: C₉H₁₂BNO₃
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425959
CAS-Nummer: 874219-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR