tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(Hydroxymethyl)piperidine, N-BOC protected

H52784.06 H52784.14
H52784.06EA 117 EUR
H52784.06 H52784.14
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate

Purity: 97% CAS No.: 116574-71-1 Molecular Formula: C11H21NO3 Formula Weight: 215.29 Melting Point: 77-81° MDL No.: MFCD03094733

Formel: C₁₁H₂₁NO₃
Molecular Weight: 215.29 g/mol
Schmelzpunkt: 77…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094733
CAS-Nummer: 116574-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR