(S)-1-Boc-2-benzylpiperazine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-tert-Butyl 2-benzylpiperazine-1-carboxylate

H52785.03
H52785.03EA 276 EUR
H52785.03
(S)-1-Boc-2-benzylpiperazine 97%
(S)-1-Boc-2-benzylpiperazine

Purity: 97% CAS No.: 169447-86-3 Molecular Formula: C16H24N2O2 Formula Weight: 276.38 MDL No.: MFCD03787923

Formel: C₁₆H₂₄N₂O₂
Molecular Weight: 276,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787923
CAS-Nummer: 169447-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR