2-(Trifluoromethyl)pyrimidine-4-carboxamide 97%

Lieferant: Thermo Scientific
Achtung

H52803.03
H52803.03EA 208 EUR
H52803.03
2-(Trifluoromethyl)pyrimidine-4-carboxamide 97%
2-(Trifluoromethyl)pyrimidine-4-carboxamide

Purity: 97% CAS No.: 914348-10-0 Molecular Formula: C6H4F3N3O Formula Weight: 191.11 MDL No.: MFCD05662689

Formel: C₆H₄F₃N₃O
Molecular Weight: 191,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662689
CAS-Nummer: 914348-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR