Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate 97%

Lieferant: Thermo Fisher Scientific
Achtung

H52818.03
H52818.03EA 499 EUR
H52818.03
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate 97%
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate

Purity: 97% CAS No.: 878745-51-8 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.12 MDL No.: MFCD05662690

Formel: C₇H₅F₃N₂O₂
Molecular Weight: 206.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662690
CAS-Nummer: 878745-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR