(S)-3-(Aminomethyl)-1-Boc-piperidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

H52822.03 H52822.06
H52822.03EA 112 EUR
H52822.03 H52822.06
(S)-3-(Aminomethyl)-1-Boc-piperidin 97%
(S)-3-(Aminomethyl)-1-Boc-piperidin

Purity: 97% CAS No.: 140645-24-5 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 MDL No.: MFCD03839877

Formel: C₁₁H₂₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03839877
CAS-Nummer: 140645-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR