Boc-O-methyl-L-serine 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-2-amino-3-methoxypropionic acid, Boc-Ser(Me)-OH, N-tert-Butoxycarbonyl-O-methyl-L-serine, (2S)-2-Amino-3-methoxypropanoic acid, N-BOC protected

H52829.MD H52829.03
H52829.MDEA 68.9 EUR
H52829.MD H52829.03
Boc-O-methyl-L-serine 97%
Boc-O-methyl-L-serine

Purity: 97% CAS No.: 51293-47-1 Molecular Formula: C9H17NO5 Formula Weight: 219.24 MDL No.: MFCD00153314

Formel: C₉H₁₇NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00153314
CAS-Nummer: 51293-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR