4-Methoxycarbonyl-2-nitrophenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Methoxycarbonyl)-2-nitrophenylboronic acid, 4-(Methoxycarbonyl)-2-nitrobenzeneboronic acid, 4-Methoxycarbonyl-2-nitrobenzeneboronic acid

H52863.06
H52863.06EA 81 EUR
H52863.06
4-Methoxycarbonyl-2-nitrophenylboronsäure ≥97%
4-Methoxycarbonyl-2-nitrophenylboronsäure

Purity: 97% CAS No.: 85107-55-7 Molecular Formula: C8H8BNO6 Formula Weight: 224.96 Melting Point: 160-171° MDL No.: MFCD01632202

Formel: C₈H₈BNO₆
Molecular Weight: 224,96 g/mol
Schmelzpunkt: 160…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632202
CAS-Nummer: 85107-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR