3-Chlor-4-(cyclopropylcarbamoyl)phenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3-Chloro-4-(cyclopropylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid

H52910.MD
H52910.MDEA 98.8 EUR
H52910.MD
3-Chlor-4-(cyclopropylcarbamoyl)phenylboronsäure ≥97%
3-Chlor-4-(cyclopropylcarbamoyl)phenylboronsäure

Purity: 97% CAS No.: 850589-44-5 Molecular Formula: C10H11BClNO3 Formula Weight: 239.46 Melting Point: 216-220° MDL No.: MFCD07363770

Formel: C₁₀H₁₁BClNO₃
Molecular Weight: 239,46 g/mol
Schmelzpunkt: 216…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363770
CAS-Nummer: 850589-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR