3-(Methylsulfonyl)phenylboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H52916.03 H52916.06
H52916.03EA 143 EUR
H52916.03 H52916.06
3-(Methylsulfonyl)phenylboronsäure ≥98%
3-(Methylsulfonyl)phenylboronsäure

Purity: 98% CAS No.: 373384-18-0 Molecular Formula: C7H9BO4S Formula Weight: 200.02 Melting Point: 102-110° MDL No.: MFCD03092935

Formel: C₇H₉BO₄S
Schmelzpunkt: 102…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03092935
CAS-Nummer: 373384-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR