4-(Isopropylcarbamoyl)phenylboronsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(N-Isopropylaminocarbonyl)phenylboronic acid, (4-(Isopropylcarbamoyl)phenyl)boronic acid, 4-(Isopropylcarbamoyl)benzeneboronic acid

H52925.MD H52925.03
H52925.MDEA 53.7 EUR
H52925.MD H52925.03
4-(Isopropylcarbamoyl)phenylboronsäure 95%
4-(Isopropylcarbamoyl)phenylboronsäure

Purity: 95% CAS No.: 397843-67-3 Molecular Formula: C10H14BNO3 Formula Weight: 207.04 Melting Point: 162-167° MDL No.: MFCD04039353

Formel: C₁₀H₁₄BNO₃
Schmelzpunkt: 162…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039353
CAS-Nummer: 397843-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR