4-(Diethylcarbamoyl)phenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(Diethylcarbamoyl)benzeneboronic acid, 4-(N,N-Diethylaminocarbonyl)phenylboronic acid
H52986.06
H52986.06EA
142
EUR
H52986.06
4-(Diethylcarbamoyl)phenylboronsäure ≥98%
4-(Diethylcarbamoyl)phenylboronsäure
Purity: 98%
CAS No.: 389621-80-1
Molecular Formula: C11H16BNO3
Formula Weight: 221.06
Melting Point: 138-142°
MDL No.: MFCD03411949
Formel:
C₁₁H₁₆BNO₃ Molecular Weight: 221,06 g/mol Schmelzpunkt: 138…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411949 CAS-Nummer: 389621-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...