3-Ethoxy-5-(trifluormethyl)phenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Ethoxy-5-trifluoromethylphenylboronic acid

H52989.03
H52989.03EA 264 EUR
H52989.03
3-Ethoxy-5-(trifluormethyl)phenylboronsäure ≥98%
3-Ethoxy-5-(trifluormethyl)phenylboronsäure

Purity: 98% CAS No.: 871332-96-6 Molecular Formula: C9H10BF3O3 Formula Weight: 233.98 Melting Point: 173-176° MDL No.: MFCD08056361

Formel: C₉H₁₀BF₃O₃
Molecular Weight: 233,98 g/mol
Schmelzpunkt: 173…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08056361
CAS-Nummer: 871332-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR