5-Carbamoyl-2-fluorphenylboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (5-Carbamoyl-2-fluorophenyl)boronic acid, 5-Carbamoyl-2-fluorophenyboronic acid, 5-Carbamoyl-2-fluorobenzeneboronic acid

H53010.MD
H53010.MDEA 43.3 EUR
H53010.MD
5-Carbamoyl-2-fluorphenylboronsäure 95%
5-Carbamoyl-2-fluorphenylboronsäure

Purity: 95% CAS No.: 874289-39-1 Molecular Formula: C7H7BFNO3 Formula Weight: 182.95 Melting Point: 192-194° MDL No.: MFCD08436055

Formel: C₇H₇BFNO₃
Molecular Weight: 182,95 g/mol
Schmelzpunkt: 192…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08436055
CAS-Nummer: 874289-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR