4-(Methoxycarbonylamino)benzeneboronic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-((Methoxycarbonyl)amino)phenyl)boronic acid

H53027.03
H53027.03EA 292 EUR
H53027.03
4-(Methoxycarbonylamino)benzeneboronic acid 97%
4-(Methoxycarbonylamino)benzeneboronic acid

Purity: 97% CAS No.: 850567-96-3 Molecular Formula: C8H10BNO4 Formula Weight: 194.98 Melting Point: 234-236° MDL No.: MFCD03788425

Formel: C₈H₁₀BNO₄
Molecular Weight: 194,98 g/mol
Schmelzpunkt: 234…236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788425
CAS-Nummer: 850567-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR