4-(Heptyloxy)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Heptyloxyphenylboronic acid, 4-n-Heptyloxybenzeneboronic acid, 4-Heptyloxybenzeneboronic acid

H53066.06
H53066.06EA 130 EUR
H53066.06
4-(Heptyloxy)phenylboronsäure ≥97%
4-(Heptyloxy)phenylboronsäure

Purity: 97% CAS No.: 136370-19-9 Molecular Formula: C13H21BO3 Formula Weight: 236.12 MDL No.: MFCD04039169

Formel: C₁₃H₂₁BO₃
Molecular Weight: 236.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039169
CAS-Nummer: 136370-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR