4-Benzyloxy-3-methylphenylboronsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-(Benzyloxy)-3-methylphenyl)boronic acid, 4-Benzyloxy-3-methylbenzeneboronic acid

H53073.06
H53073.06EA 519 EUR
H53073.06
4-Benzyloxy-3-methylphenylboronsäure 96%
4-Benzyloxy-3-methylphenylboronsäure

Purity: 96% CAS No.: 338454-30-1 Molecular Formula: C14H15BO3 Formula Weight: 242.08 Melting Point: 102-106° MDL No.: MFCD06409944

Formel: C₁₄H₁₅BO₃
Molecular Weight: 242.08 g/mol
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06409944
CAS-Nummer: 338454-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR