3-Fluor-(5-methoxycarbonyl)phenylboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Fluoro-5-methoxycarbonylphenylboronic Acid, 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

H53084.03
H53084.03EA 589 EUR
H53084.03
3-Fluor-(5-methoxycarbonyl)phenylboronsäure 95%
3-Fluor-(5-methoxycarbonyl)phenylboronsäure

Purity: 95% CAS No.: 871329-62-3 Molecular Formula: C8H8BFO4 Formula Weight: 197.96 Melting Point: 138-142° MDL No.: MFCD07363749

Formel: C₈H₈BFO₄
Molecular Weight: 197,96 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363749
CAS-Nummer: 871329-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR