5-Chlor-2-fluorphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dihydroxy(5-chlor-2-fluorphenyl)boran, 5-Chlor-2-fluorbenzolboronsäure, (5-Chlor-2-fluorphenyl)metaborsäure

H53108.03
H53108.03EA 29.8 EUR
H53108.03
5-Chlor-2-fluorphenylboronsäure ≥97%
5-Chlor-2-fluorphenylboronsäure

Purity: 97% CAS No.: 352535-83-2 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 Melting Point: 182-188° MDL No.: MFCD05664225

Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
Schmelzpunkt: 182…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664225
CAS-Nummer: 352535-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR