4-Isopropyl-5-pyrimidinboronsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
H53118.MD
H53118.MDEA
331
EUR
H53118.MD
4-Isopropyl-5-pyrimidinboronsäure 95%
4-Isopropyl-5-pyrimidinboronsäure
Purity: 95%
CAS No.: 913835-27-5
Molecular Formula: C7H11BN2O2
Formula Weight: 165.99
Storage Temperature: 2°C to 8°C
Melting Point: 104-109°
MDL No.: MFCD08235084
Formel:
C₇H₁₁BN₂O₂ Molecular Weight: 165,99 g/mol Schmelzpunkt: 104…109 °C |
MDL:
MFCD08235084 CAS-Nummer: 913835-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...