5-(Methylsulphonyl)pyridine-3-boronic acid 98%

Lieferant: Thermo Scientific
Achtung

H53133.MD
H53133.MDEA 178 EUR
H53133.MD
5-(Methylsulphonyl)pyridine-3-boronic acid 98%
5-(Methylsulphonyl)pyridine-3-boronic acid

Purity: 98% CAS No.: 913836-01-8 Molecular Formula: C6H8BNO4S Formula Weight: 201.00 Storage Temperature: 2°C to 8°C Melting Point: 123-125° MDL No.: MFCD08689536

Formel: C₆H₈BNO₄S
Molecular Weight: 201,00 g/mol
Schmelzpunkt: 123…125 °C
MDL: MFCD08689536
CAS-Nummer: 913836-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR