1-Methyl-1H-pyrazol-4-boronsäurepinakolester ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methylpyrazole-4-boronic acid pinacol ester, 1-Methyl-4-pyrazole boronic acid pinacol ester, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

H53139.03
H53139.03EA 78 EUR
H53139.03
1-Methyl-1H-pyrazol-4-boronsäurepinakolester ≥97%
1-Methyl-1H-pyrazol-4-boronsäurepinakolester

Purity: 97% CAS No.: 761446-44-0 Molecular Formula: C10H17BN2O2 Formula Weight: 208.07 Melting Point: 59-64° MDL No.: MFCD03789259

Formel: C₁₀H₁₇BN₂O₂
Molecular Weight: 208,07 g/mol
Schmelzpunkt: 59…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03789259
CAS-Nummer: 761446-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR