[4-(1-Cyano-1-methylethyl)phenyl]boronic acid 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(2-Cyanoprop-2-yl)benzeneboronic acid, 2-(4-Boronophenyl)-2-methylpropanenitrile

H53173.MD
H53173.MDEA 464 EUR
H53173.MD
[4-(1-Cyano-1-methylethyl)phenyl]boronic acid 97%
[4-(1-Cyano-1-methylethyl)phenyl]boronic acid

Purity: 97% CAS No.: 850568-67-1 Molecular Formula: C10H12BNO2 Formula Weight: 189.02 Melting Point: 146-148° MDL No.: MFCD06659832

Formel: C₁₀H₁₂BNO₂
Molecular Weight: 189,02 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659832
CAS-Nummer: 850568-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR