3-Methyl-2-thiophenboronsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H53201.MD H53201.03
H53201.MDEA 65.2 EUR
H53201.MD H53201.03
3-Methyl-2-thiophenboronsäure ≥98%
3-Methyl-2-thiopheneboronic acid

Purity: 98% CAS No.: 177735-09-0 Molecular Formula: C5H7BO2S Formula Weight: 142.01 Melting Point: 192-196° MDL No.: MFCD00236049

Formel: C₅H₇BO₂S
Schmelzpunkt: 192…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236049
CAS-Nummer: 177735-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR