1-Phenylvinylboronic acid pinacol ester 95%
Lieferant: Thermo Scientific
Synonyme:
4,4,5,5-Tetramethyl-2-(1-phenylvinyl)-1,3,2-dioxaborolane
H53216.03
H53216.03EA
75
EUR
H53216.03
1-Phenylvinylboronic acid pinacol ester 95%
1-Phenylvinylboronic acid pinacol ester
Purity: 95%
CAS No.: 143825-84-7
Molecular Formula: C14H19BO2
Formula Weight: 230.11
Melting Point: 40-42°
MDL No.: MFCD06659923
Formel:
C₁₄H₁₉BO₂ Molecular Weight: 230,11 g/mol Schmelzpunkt: 40…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659923 CAS-Nummer: 143825-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...