4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(4-(2-Bromoethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H53246.03
H53246.03EA 135 EUR
H53246.03
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 97%
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester

Purity: 98% CAS No.: 913836-27-8 Molecular Formula: C14H20BBrO3 Formula Weight: 327.03 Melting Point: 90-92° MDL No.: MFCD09027285

Formel: C₁₄H₂₀BBrO₃
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027285
CAS-Nummer: 913836-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR