2-Formyl-4-thiophenboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Formylthiophene-4-boronic acid

H53298.03
H53298.03EA 82.6 EUR
H53298.03
2-Formyl-4-thiophenboronsäure ≥98%
2-Formyl-4-thiophenboronsäure

Purity: 98% CAS No.: 175592-59-3 Molecular Formula: C5H5BO3S Formula Weight: 156.97 Storage Temperature: 2°C to 8°C Melting Point: 113° MDL No.: MFCD03002365

Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
Schmelzpunkt: 113 °C
MDL: MFCD03002365
CAS-Nummer: 175592-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR