2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester 95%

Lieferant: Thermo Scientific

H53309.03
H53309.03EA 104 EUR
H53309.03
2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester 95%
2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester

Purity: 95% CAS No.: 441011-76-3 Molecular Formula: C12H16BBrO2 Formula Weight: 282.97 MDL No.: MFCD04039022

Formel: C₁₂H₁₆BBrO₂
Molecular Weight: 282,97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039022
CAS-Nummer: 441011-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR