2-(Hydroxymethyl)pyridine-5-boronic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-(Hydroxymethyl)pyridine-3-boronic acid

H53319.MD
H53319.MDEA 331 EUR
H53319.MD
2-(Hydroxymethyl)pyridine-5-boronic acid 97%
2-(Hydroxymethyl)pyridine-5-boronic acid

Purity: 97% CAS No.: 913835-98-0 Molecular Formula: C6H8BNO3 Formula Weight: 152.94 Storage Temperature: 2°C to 8°C MDL No.: MFCD09027230

Formel: C₆H₈BNO₃
Molecular Weight: 152,94 g/mol
MDL: MFCD09027230
CAS-Nummer: 913835-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR